missing translation for 'onlineSavingsMsg'
Learn More

BFH772, MedChemExpress

Product Code. p-200097521
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16445747 5 mg 5mg
16444316 100 mg 100mg
16443008 10 mg 10mg
16439576 50 mg 50mg
16402586 25 mg 25mg
5 options
This item is not returnable. View return policy

Product Code. 16445747

Brand: Medchem Express HY100419

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BFH772 is a potent oral VEGFR2 inhibitor, which is highly effective at targeting VEGFR2 kinase with an IC50 value of 3 nM.

BFH772 is a potent oral VEGFR2 inhibitor, which is highly effective at targeting VEGFR2 kinase with an IC50 value of 3 nM.

Specifications

Chemical Name or Material BFH772
CAS 890128-81-1
Color White
Purity Grade Notes Research
Molecular Formula C23H16F3N3O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (227.59 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(F)(F)F)=CC=C1)C2=CC=CC3=CC(OC4=NC=NC(CO)=C4)=CC=C32
Molecular Weight (g/mol) 439.39
Formula Weight 439.39
Percent Purity 96.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.