Learn More
BFH772, MedChemExpress
Product Code. 16445747
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16445747
Brand: Medchem Express HY100419
MedChemExpress BFH772 is a potent oral VEGFR2 inhibitor, which is highly effective at targeting VEGFR2 kinase with an IC50 value of 3 nM.
BFH772 is a potent oral VEGFR2 inhibitor, which is highly effective at targeting VEGFR2 kinase with an IC50 value of 3 nM.
Specifications
Chemical Name or Material BFH772
CAS 890128-81-1
Color White
Purity Grade Notes Research
Molecular Formula C23H16F3N3O3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (227.59 mM; Need ultrasonic)
SMILES O=C(NC1=CC(C(F)(F)F)=CC=C1)C2=CC=CC3=CC(OC4=NC=NC(CO)=C4)=CC=C32
Molecular Weight (g/mol) 439.39
Formula Weight 439.39
Percent Purity 96.25%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.