Learn More
Bevirimat, TRC
Product Code. 17989574
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17989574 2.5 mg 2.5mg
17978524 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17989574 Supplier Toronto Research Chemicals Supplier No. TRCB3347002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 174022-42-5
Molecular Formula C36 H56 O6
Molecular Weight (g/mol) 584.83
Synonym 3-O-(3',3'-Dimethylsuccinyl)betulinic acid, Bevirimat, FH 11327, O-(3,3-Dimethylsuccinyl)betulinic Acid, PA 457, YK FH312
IUPAC Name (3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-9-(3-carboxy-3-methyl-butanoyl)oxy-1-isopropenyl-5a,5b,8,8,11a-pentamethyl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysene-3a-carboxylic acid
SMILES CC(=C)C1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](OC(=O)CC(C)(C)C(=O)O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(=O)O
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C36H56O6/c1-21(2)22-12-17-36(30(40)41)19-18-34(8)23(28(22)36)10-11-25-33(7)15-14-26(42-27(37)20-31(3,4)29(38)39)32(5,6)24(33)13-16-35(25,34)9/h22-26,28H,1,10-20H2,2-9H3,(H,38,39)(H,40,41)/t22?,23-,24+,25-,26+,28-,33+,34-,35-,36+/m1/s1
Formula Weight 584.4077
Chemical Name or Material Bevirimat