Learn More
Bevirimat, MedChemExpress
Product Code. 16491528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491528 5 mg 5mg
16449128 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16491528 Supplier Medchem Express Supplier No. HYN0842
MedChemExpress Bevirimat (PA-457; MPC-4326; YK FH312) is an anti-HIV drug derived from a betulinic acid-like compound; is believed to inhibit HIV by a novel mechanism, so-called maturation inhibition.
Bevirimat (PA-457
Specifications
Chemical Name or Material Bevirimat
Physical Form Solid
CAS 174022-42-5
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C36H56O6
For Use With (Application) COVID-19-anti-virus
Synonym PA-457 MPC-4326 YK FH312
Solubility Information DMSO : ≥ 50 mg/mL (85.49 mM)
SMILES OC([C@]1(CC[C@H]2C(C)=C)[C@@]2([H])[C@](CC[C@@]3([H])[C@]4(CC[C@]5([H])[C@@]3(CC[C@H](OC(CC(C)(C)C(O)=O)=O)C5(C)C)C)C)([H])[C@@]4(C)CC1)=O
Molecular Weight (g/mol) 584.83
Formula Weight 584.83
Percent Purity 98.0%
Grade Research