Learn More
Buerkle™ Power Supply Unit for Buerkle™ AccuOne/EnergyOne
Product Code. 11961641 Shop All Bürkle Products
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
11961641 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11961641 Supplier Buerkle™ Supplier No. 59003020
Estimated Shipment: 02-12-2026
Includes an international plug adapter
  • Power supply via 230 V power supply unit
  • International plug adapter included with delivery
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Accu/EnergyOne Barrel Pump
Voltage 230 V
Description Power supply unit incl. Connectors for Accu/EnergyOne
Includes EU, UK, US, AU plug
Weight (Metric) 0.01 kg
Product Type Power Supply Unit