Learn More
Buerkle™ Hose Angle Bracket
Provided with cap nut, inner thread and reducing nozzle.
Brand: Buerkle™ 8548-0020
57.44 NOK valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 0.02100kg
Description
- Angled at 90°
- Manufactured from polypropylene
Specifications
Hose Angle Bracket | |
Cap nut, inner thread and reducing nozzle |