Learn More
Bethanechol chloride, MedChemExpress
Product Code. 16421416
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
5 g
Unit Size:
200mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421416 200 mg 200mg
16420918 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16421416 Supplier Medchem Express Supplier No. HYB0406A
MedChemExpress Bethanechol chloride (Carbamyl-β-methylcholine chloride), a parasympathomimetic agent, is a mAChR agonist that exerts its effects via directly stimulating the mAChR (M1, M2, M3, M4, and M5) of the parasympathetic nervous system.
Bethanechol chloride (Carbamyl-β-methylcholine chloride), a parasympathomimetic agent, is a mAChR agonist that exerts its effects via directly stimulating the mAChR (M1, M2, M3, M4, and M5) of the parasympathetic nervous system.
Especificaciones
Chemical Name or Material Bethanechol chloride
Physical Form Solid
CAS 590-63-6
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C7H17ClN2O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym Carbamyl-β-methylcholine chloride
Solubility Information H2O : ≥ 50 mg/mL (254.22 mM) ∣DMSO : 11.11 mg/mL (56.49 mM; Need ultrasonic)
SMILES CC(OC(N)=O)C[N+](C)(C)C.[Cl-]
Molecular Weight (g/mol) 196.68
Formula Weight 196.68
Percent Purity 95.0%
Grade Research
Mostrar más Mostrar menos
Research purposes only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.