missing translation for 'onlineSavingsMsg'
Learn More

Bethanechol Chloride, 100.2%

Product Code. 11404622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404622 25 g 25g
11494612 5 g 5g
11414622 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404622 Supplier MP Biomedicals Supplier No. 0219016025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 590-63-6
Molecular Formula C7H17ClN2O2
Molecular Weight (g/mol) 196.675
InChI Key XXRMYXBSBOVVBH-UHFFFAOYSA-N
Synonym bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone
PubChem CID 11548
ChEBI CHEBI:3085
IUPAC Name 2-carbamoyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)N.[Cl-]

Specifications

Color White
Assay Percent Range 100.2%
Quantity 25 g
Formula Weight 196.7
Physical Form Powder
Chemical Name or Material Bethanechol Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.