Learn More
Thermo Scientific™ BetaSil™ Silica Guard Cartridges
Product Code. 10514855 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric):
1 mm I.D.
2.1 mm
3 mm
4 mm
Length (Metric):
10 mm
Particle Size:
5 μm
Unit Size:
Pack of 4
Product Code. Diameter (Metric) Length (Metric) Particle Size unitSize
10514855 1 mm I.D. 10 mm 5 μm Pack of 4
10187644 2.1 mm 10 mm 5 μm Pack of 4
10279174 3 mm 10 mm 5 μm Pack of 4
10239224 4 mm 10 mm 5 μm Pack of 4
4 options
Product Code. 10514855
Brand: Thermo Scientific™ 70005011001
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
  • 100&Angst; silica
  • High surface area
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) 1 mm I.D.
Diameter (Metric) Inner 1 mm
Product Line BetaSil