Learn More
Thermo Scientific™ BetaSil™ Diol Guard Cartridge
Product Code. 10095814 Shop All Thermo Scientific Products
Change view
Click to view available options
Particle Size:
5 μm
Diameter (Metric):
4 mm
Length (Metric):
10 mm
Unit Size:
Pack of 4
Product Code. Particle Size Diameter (Metric) Length (Metric) unitSize
10095814 5 μm 4 mm 10 mm Pack of 4
1 options
Product Code. 10095814
Brand: Thermo Scientific™ 72605014001
Only null left
Add to basket
Add to basket
The Thermo Scientific™ BetaSil™ Diol Guard Cartridge is available in limited dimensions. Please contact Customer Service for more details.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 5 μm
Length (Metric) 10 mm
Diameter (Metric) 4 mm
Description Particle Size: 5um; 10L x 4.0mm I.D.
For Use With (Application) LC/MS
Product Line Beta