Learn More
Texwipe™ BetaMop™ II
Product Code. p-209838 Shop All Texwipe Products
Click to view available options
Product Type:
BetaMop II Mop
Refill Mop Head
For Use With (Equipment):
BetaMop II
Wring-bucket Systems
Unit Size:
Each
Pack of 6
Product Code. 11581214
Brand: Texwipe™ TX7106
Only null left
Add to basket
Add to basket
Low contamination properties make this the perfect mop for floor cleaning in critical environments
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Product Type BetaMop II Mop
Autoclavable Autoclavable
Resistant To Corrosion
Includes Handle and Head Refill
For Use With (Equipment) Wring-bucket Systems
For Use With (Application) For floor cleaning in both electronic cleanrooms and sterile environments
Material Polyester