Description
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Specifications
| Product Type | Refill Mop Head |
| Autoclavable | Autoclavable |
| Resistant To | Corrosion |
| For Use With (Equipment) | BetaMop II |
| For Use With (Application) | For Floor Cleaning in Critical Environments |
| Material | Polyester |