missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Amyloid beta (aa1-42) Antibody, R&D Systems™

Mouse Monoclonal Antibody
1950.00 NOK - 4345.00 NOK
Specifications
| Antigen | betaAmyloid |
|---|---|
| Clone | MM26 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724498
![]() |
R&D Systems
MAB9618-SP |
25 μg |
1950.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18790118
![]() |
R&D Systems
MAB9618-100 |
100 μg |
4345.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta Amyloid Monoclonal specifically detects beta Amyloid in Human samples. It is validated for ELISA.Specifications
| betaAmyloid | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Abeta, Amyloid beta, beta Amyloid, beta-Amyloid Peptide, Protease Nexin II | |
| Recombinant protein to human Amyloid beta 1-42 peptide | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Amyloid beta (aa1-42) in sandwich ELISAs. |
| MM26 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 351 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title