missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt, MP Biomedicals
Shop All MP Biomedicals ProductsSpecifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 709.4 |
| Chemical Name or Material | Beta-Nicotinamide Adenine Dinucleotide Reduced Disodium Salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?