missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (rTUBB3/7406), Novus Biologicals™
Mouse Monoclonal Antibody
2300.00 NOK - 4570.00 NOK
Specifications
| Antigen | beta-III Tubulin |
|---|---|
| Clone | rTUBB3/7406 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635104
|
Novus Biologicals
NBP3-20486-100ug |
100 μg |
4570.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690295
|
Novus Biologicals
NBP3-20486-20ug |
20 μg |
2300.00 NOK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| beta-III Tubulin | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific | |
| 10mM PBS with 0.05% BSA | |
| 10381 | |
| IgG1 κ | |
| Protein A or G purified |
| rTUBB3/7406 | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Recombinant fragment (around aa250-450) of human beta-III Tubulin (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title