missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-III Tubulin Antibody (CL5813), Novus Biologicals™
Mouse Monoclonal Antibody
3370.00 NOK - 4805.00 NOK
Specifications
| Antigen | beta-III Tubulin |
|---|---|
| Clone | CL5813 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256654
|
Novus Biologicals
NBP2-61430 |
100 μL |
4805.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646768
|
Novus Biologicals
NBP2-61430-25ul |
25 μL |
3370.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| beta-III Tubulin | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| 10381 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: YEDDEEESEAQGPK | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL5813 | |
| Monoclonal | |
| Purified | |
| Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Neuroscience | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| TUBB3 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title