Learn More
beta-Hydroxyisovaleric Acid, TRC
Product Code. 30134385
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134385 50 g 50g
30132731 1 g 1g
30008187 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134385 Supplier Toronto Research Chemicals Supplier No. TRCH94360550G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 625-08-1
Molecular Formula C5 H10 O3
Molecular Weight (g/mol) 118.13
Synonym Butanoic acid, 3-hydroxy-3-methyl-, Butyric acid, 3-hydroxy-3-methyl- (6CI,7CI,8CI), 3-Hydroxy-3-methylbutanoic acid, 3-Hydroxy-3-methylbutyric acid, 3-Hydroxyisovaleric acid, 3-Methyl-3-hydroxybutyric acid, β-Hydroxy-β-methylbutyric acid, β-Hydroxyisovaleric acid
IUPAC Name 3-hydroxy-3-methylbutanoic acid
SMILES CC(C)(O)CC(=O)O
Specifications
Quantity 50 g
InChI Formula InChI=1S/C5H10O3/c1-5(2,8)3-4(6)7/h8H,3H2,1-2H3,(H,6,7)
Formula Weight 118.063
Chemical Name or Material beta-Hydroxyisovaleric Acid