Learn More
Fisherbrand™ Beta/Gamma Protection Duoshield Box
Product Code. 12327197 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W x H):
L x W : 45 x 30 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12327197 L x W : 45 x 30 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12327197
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
Provides radiation protection shielding for use in laboratories.
  • Transparent shielding
  • Constructed of acrylic provides easy fabrication and manufacturability
Specifications
Color Transparent
Dimensions (L x W x H) L x W : 45 x 30 cm
For Use With (Application) beta/gamma radiation protection
Includes Curved base
Product Type Duoshield box
Material Acrylic