Learn More
Beta Arrestin 1 Mouse anti-Human, Unconjugated, Clone: 1H8E4, Proteintech
Shop All Proteintech ProductsDescription
The Beta Arrestin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Arrestin 1. This antibody recognizes human, mouse, rat antigen. The Beta Arrestin 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
| Antigen | Beta Arrestin 1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1H8E4 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 408 |
| Show More |