Learn More
Beta Arrestin 1 Mouse anti-Human, Unconjugated, Clone: 1H8E4, Proteintech
Product Code. 16800296 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16800296 20 μL 20µL
16890286 150 μL 150µL
2 options
Product Code. 16800296
Brand: Proteintech 675801IG20UL
Mouse Monoclonal Antibody
The Beta Arrestin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Arrestin 1. This antibody recognizes human, mouse, rat antigen. The Beta Arrestin 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen Beta Arrestin 1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1H8E4
Concentration 1000 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 408
Target Species Human, Rat, Mouse
Isotype IgG2a