Learn More
Bestatin, MedChemExpress
Product Code. 16484247
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484247 50 mg 50mg
16468358 10 mg 10mg
16438126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16484247 Supplier Medchem Express Supplier No. HYB0134
MedChemExpress Bestatin is a natural, broad-spectrum, and competitive CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase inhibitor. Bestatin has anticancer effects.
Bestatin is a natural, broad-spectrum, and competitive CD13 (Aminopeptidase N)/APN and leukotriene A4 hydrolase inhibitor. Bestatin has anticancer effects.
Specifications
Chemical Name or Material Bestatin
Physical Form Solid
CAS 58970-76-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C16H24N2O4
For Use With (Application) Cancer-Kinase/protease
Synonym Ubenimex
Solubility Information DMSO : 8.33 mg/mL (27.01 mM; Need ultrasonic)
SMILES CC(C)C[C@@H](C(O)=O)NC([C@@H](O)[C@H](N)CC1=CC=CC=C1)=O
Molecular Weight (g/mol) 308.37
Formula Weight 308.37
Percent Purity 95.0%
Grade Research