missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BES, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Descripción
BES is used as a buffer to maintain the pH of the solutions in biological experiments. It is useful for diagnostic assay manufacturing industry. It induces cross-linking between sulfonated polyimide chains in sulfonated polyimide membranes. It acts as biobuffer to investigate the aqueous medium self-assembly of heterometallic CuII/Li 3D coordination polymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Especificaciones
Melting Point | 153°C to 157°C |
Quantity | 25 g |
Beilstein | 1781572 |
Solubility Information | Soluble in water. |
Formula Weight | 213.25 |
Percent Purity | 99% |
Chemical Name or Material | BES |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'