Learn More
Berkshire™ Knitted Polyester Lab-Tips Swabs
Shop All Berkshire Products
Click to view available options
Tip Style:
Flexible
Rigid
Length (Metric) Handle:
59 mm
100 mm
Length (Metric) Tip:
2.5 mm
10.9 mm
Unit Size:
Pack of 2000
Pack of 500
Description
- Continuous filament polyester knit head
- Short polypropylene handle with a rigid head
- Thermally bonded head
- Laundered and packaged in Class 10 (ISO 4) cleanroom
- Autoclavable
Specifications
Specifications
| Certifications/Compliance | ISO Class 4 |
| Diameter (English) Handle | 0.08 in. |
| Length (English) | 2.8 in. |
| For Use With (Application) | Designed for the highest level of contamination control in critical processing applications, cleaning of sensitive equipment, applying lubricants and other liquids, removing excess materials |
| Handle Color | Green |
| Product Type | Swab |
| Diameter (Metric) Handle | 2.2 mm |
| Length (Metric) | 70 mm |
| Length (English) Handle | 2.32 in. |
| Length (Metric) Handle | 59 mm |
| Show More |