Learn More
Berkshire™ Durx 570 Wiper
Product Code. 18801375 Shop All Berkshire Products
Change view
Click to view available options
Dimensions (L x W):
22.86 x 22.86 cm (9 x 9 in.)
Unit Size:
Pack of 6000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
18801375 22.86 x 22.86 cm (9 x 9 in.) Pack of 6000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18801375 Supplier Berkshire™ Supplier No. DR570.0909.20
  • ONLINE EXCLUSIVE PRICE
    7663.76 NOK
    6074.09 NOK / Pack of 6000
    Save 1589.67 NOK
    20% Off
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
For use in ISO Class 5 to 8 controlled environments
  • Autoclavable
  • Free of chemical additives
Specifications
Product Type Cleanroom Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Non-woven Polyester Cellulose
Dimensions (L x W) 22.86 x 22.86 cm (9 x 9 in.)
For Use With (Application) Designed for use in ISO Class 5 and higher cleanroom environments and USP <797> applications