Description
- Smooth, highly absorbent and durable with low extractable levels
- Economical
- Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
- 45% polyester and 55% cellulose
- Each package is gamma irradiated to a 10-6 Sterility Assurance
Specifications
Specifications
| Length (English) | 9 in. |
| Length (Metric) | 23 cm |
| Width (English) | 9 in. |
| Width (Metric) | 23 cm |
| Material | Non-woven Polyester Cellulose |
| Color | White |
| Dimensions (L x W) | 9 x 9 in. |
| For Use With (Application) | Designed for use in ISO Class 5 and higher sterile environments and USP <797> applications |
| Sterility | Sterile |