Learn More
Beperidium iodide, MedChemExpress
Product Code. 16422808
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422808 10 mg 10mg
16465547 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16422808 Supplier Medchem Express Supplier No. HY100152
MedChemExpress Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Beperidium iodide shows a competitive antagonistic effect against acetylcholine receptor with a pA2 of 7.93.
Specifications
Chemical Name or Material Beperidium iodide
CAS 86434-57-3
Color White
Purity Grade Notes Research
Molecular Formula C23H34IN3O3
Quantity 10 mg
Synonym SX 810
Solubility Information DMSO : ≥ 100 mg/mL (189.60 mM)
SMILES C[N+]1(CC)CCC(OC(C(C2=NOC3=C2C=CC=C3)N4CCCCCC4)=O)CC1.[I-]
Molecular Weight (g/mol) 527.44
Formula Weight 527.44
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation