missing translation for 'onlineSavingsMsg'
Learn More

Benzyloxyacetaldehyde diethyl acetal, 98%

Product Code. 11411808
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411808 50 g 50g
11401808 10 g 10g
11421808 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411808 Supplier Thermo Scientific Alfa Aesar Supplier No. A14474.18
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 42783-78-8 | C13H20O3 | 224.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42783-78-8
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.3
MDL Number MFCD00051786
InChI Key VTYTZCJKJNWMGA-UHFFFAOYSA-N
Synonym 2,2-diethoxyethoxy methyl benzene, benzyloxyacetaldehyde diethyl acetal, benzene, 2,2-diethoxyethoxy methyl, acmc-20alp5, 1,1-diethoxy-2-benzyloxy ethane, 2,2-diethoxy-ethoxymethyl-benzene, 2,2-diethoxy-1-phenylmethoxy ethane, 2-benzyloxy acetaldehyde diethyl acetal
PubChem CID 4321487
IUPAC Name 2,2-diethoxyethoxymethylbenzene
SMILES CCOC(COCC1=CC=CC=C1)OCC

Specifications

Density 0.987
Boiling Point 99°C to 100°C (0.6 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.479
Quantity 50 g
Beilstein 3278795
Formula Weight 224.3
Percent Purity 98%
Chemical Name or Material Benzyloxyacetaldehyde diethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.