Learn More
Benzyl methacrylate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2495-37-6 |
| Molecular Formula | C11H12O2 |
| MDL Number | MFCD00053684 |
| Quantity | 1 kg |
| SMILES | CC(C(OCC1=CC=CC=C1)=O)=C |
| Molecular Weight (g/mol) | 176.21 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzyl methacrylate |