Learn More
Benzyl Benzoate, Sigma-Aldrich™
Product Code. 15850561
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15850561 1 L 1 L
15840561 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15850561 Supplier Sigma Aldrich Supplier No. 8187011000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 120-51-4
Grade Research
Molecular Weight (g/mol) 212.24
Specifications
Melting Point 21°C
Density 1.12 g/cm3 at 20°C
Boiling Point 324°C/1013 hPa
MDL Number MFCD00003075
Quantity 1 L
Synonym Benzyl benzoate, Benzoic acid benzyl ester, Benzyl benzoate