missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, Sigma-Aldrich™

Product Code. 15881731
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Unit Size:
1 L
100mL
2.5 L
25 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15881731 25 L 25 L
15851731 100 mL 100mL
15861731 1 L 1 L
15871731 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15881731 Supplier Sigma Aldrich Supplier No. 8222599025
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-51-6
Grade Research
Molecular Formula C6H5CH2OH
Molecular Weight (g/mol) 108.14

Specifications

Melting Point −16-−13°C (Lit.)
Density 1.045 g/mL at 25°C
Boiling Point 203°C to 205°C
MDL Number MFCD00004599
Quantity 25 L
Synonym Benzenemethanol, Benzenemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.