missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, ReagentPlus™, ≥99%, Honeywell™

Product Code. 15620990 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
100mL
2.5 L
20 L
500mL
Product Code. Quantity Packaging unitSize
15620990 500 mL Glass Bottle 500mL
15680980 100 mL Glass Bottle 100mL
15690980 1 L Glass Bottle 1 L
15600990 2.5 L Glass Bottle 2.5 L
15610990 20 L Stainless Steel Can 20 L
5 options
This item is not returnable. View return policy

Product Code. 15620990

Brand: Honeywell Research Chemicals 108006500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 100-51-6
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00004599,MFCD03792087
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
Synonym benzyl alcohol, benzenemethanol, phenylcarbinol, benzoyl alcohol, hydroxytoluene, benzenecarbinol, phenylmethyl alcohol, alpha-toluenol, hydroxymethyl benzene, benzylalcohol
PubChem CID 244
ChEBI CHEBI:17987
IUPAC Name phenylmethanol
SMILES OCC1=CC=CC=C1

Especificaciones

Melting Point -15.4°C
Color Colorless
Density 1.045g/mL at 25°C
Boiling Point 205.3°C
Flash Point 96°C (204.8°F)
Ignition Point 817°F
Packaging Glass Bottle
Linear Formula C6H5CH2OH
Refractive Index n20/D 1.539(lit.)
Quantity 500 mL
UN Number UN3334
Beilstein 878307
Vapor Density 3.7 (vs air)
Formula Weight 108.14g/mol
Vapor Pressure 3.75mmHg (77°C), 13.3mmHg (100°C)
Percent Purity ≥99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material Benzyl Alcohol
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Benzyl alcohol
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Indicaciones de peligro
  • H302 + H332-Harmful if swallowed or if inhaled.
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.