Learn More
Benzyl Acetate, Sigma-Aldrich™
Product Code. 15888930
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888930 1 L 1 L
15878930 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888930 Supplier Sigma Aldrich Supplier No. 8031811000
 Restricted Item
Estimated Shipment: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 140-11-4
Grade Research
Molecular Weight (g/mol) 150.17
Specifications
Melting Point -51°C
Density 1.06 g/cm3 at 20°C
Boiling Point 205°C to 207°C/1013 hPa
MDL Number MFCD00008712
Quantity 1 L
Synonym Benzyl acetate, Acetic acid benzyl ester, Benzyl acetate