Learn More
Benzyl acetate, 99+%
Product Code. 10274700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10274700 1 L 1 L
10020550 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10274700 Supplier Thermo Scientific Acros Supplier No. 105820010
Request Bulk or Custom Format
CAS: 140-11-4 | C9H10O2 | 150.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 140-11-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
InChI Key QUKGYYKBILRGFE-UHFFFAOYSA-N
IUPAC Name benzyl acetate
SMILES CC(=O)OCC1=CC=CC=C1
Specifications
Melting Point -51.0°C
Density 1.0550 g/mL
Boiling Point 214.0°C
Flash Point 102°C
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula CH3CO2CH2C6H5
Refractive Index 1.5010 to 1.5030
Quantity 1 L
Beilstein 06, 435
Merck Index 15, 1126
Specific Gravity 1.055
Solubility Information Solubility aq. soln.: 1.5 g/L water (20°C). Other solubilities: miscible with alcohol and ether
Formula Weight 150.18
Chemical Name or Material Benzyl acetate
Show More Show Less
Product Identifier
  • Benzyl acetate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement