Learn More
Benzquinamide, LoGiCal
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Benzquinamide
Specifications
Specifications
| Chemical Name or Material | Benzquinamide |
| CAS | 63-12-7 |
| Analyte or Component Names | Benzquinamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H32 N2 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2 H-Benzo[a]quinolizine-3-carboxamide, N,N-diethyl-1,3,4,6,7,11 b-hexahydro-2-hydroxy-9,10-dimethoxy-, acetate (ester) (8 CI), 2-Hydroxy-3-diethylcarbamoyl-9,10-dimethoxy-1,2,3,4,6,7-hexahydro-11 b,4-benzoquinolizine acetate, 2-Hydroxy-3-diethylcarbamyl-9,10-dimethoxy-1,2,3,4,6,7-hexahydro-11 bH-benzoquinolizine acetate, BZQ, Benzoquinamide, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy-, 2 H-Benzo[a]quinolizine-3-carboxamide, 2-(acetyloxy)-N,N-diethyl-1,3,4,6,7,11 b-hexahydro-9,10-dimethoxy- |
| InChI Formula | InChI=1 S/C22H32N2O5/c1-6-23(7-2)22(26)17-13-24-9-8-15-10-20(27-4)21(28-5)11-16(15)18(24)12-19(17)29-14(3)25/h10-11,17-19 H,6-9,12-13H2,1-5H3 |
| SMILES | CCN(CC)C(=O)C1CN2CCc3cc(OC)c(OC)cc3C2CC1OC(=O)C |
| IUPAC Name | [3-(diethylcarbamoyl)-9,10-dimethoxy-2,3,4,6,7,11 b-hexahydro-1 H-benzo[a]quinolizin-2-yl] acetate |
| Show More |