Learn More
Benzoylmesaconine, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Benzoylmesaconine is the most abundant component of Wutou decoction, which is widely used in China because of its therapeutic effect on rheumatoid arthritis.
Specifications
Specifications
| Chemical Name or Material | Benzoylmesaconine |
| CAS | 63238-67-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H43NO10 |
| Quantity | 5 mg |
| Synonym | Mesaconine 14-benzoate |
| Solubility Information | DMSO : 100 mg/mL (169.59 mM; Need ultrasonic) |
| SMILES | CO[C@@H]1C2([C@@](C[C@@]3(O)[C@@H]4OC(C5=CC=CC=C5)=O)([H])[C@@]4([H])[C@](O)([C@@H](O)[C@@H]3OC)C6C2N(C)C7)[C@@]([C@H]6OC)([H])[C@@]7(COC)[C@H](O)C1 |
| Molecular Weight (g/mol) | 589.67 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.