Learn More
Benzoyl Chloride, Sigma-Aldrich™
Product Code. 15818660
Change view
Click to view available options
Quantity:
1 L
30 kg
Unit Size:
1 L
30kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15818660 30 kg 30kg
15808660 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15818660 Supplier Sigma Aldrich Supplier No. 8018049030
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-88-4
Grade Research
Molecular Weight (g/mol) 140.57
Specifications
Melting Point -0.6°C
Density 1.21 g/cm3 at 20°C
Boiling Point 197.2°C/1013 hPa
MDL Number MFCD00000653
Quantity 30 kg
Synonym Benzoyl chloride, Benzoic acid chloride, Benzoyl chloride