Learn More
Benzotrifluoride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 98-08-8 |
| Melting Point | -29°C |
| Density | 1.197 g/cm3 at 20°C |
| Boiling Point | 100°C to 103°C/1013 hPa |
| MDL Number | MFCD00000372 |
| Quantity | 100 mL |
| Synonym | Benzotrifluoride, Benzotrifluoride |
| Molecular Weight (g/mol) | 146.11 |
| Grade | Research |