Learn More
Benzoin, Sigma-Aldrich™
Product Code. 15878640
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878640 1 kg 1kg
15868640 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15878640 Supplier Sigma Aldrich Supplier No. 8017761000
 Restricted Item
Estimated Shipment: 30-10-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 119-53-9
Grade Research
Molecular Formula C6H5CH(OH)COC6H5
Molecular Weight (g/mol) 212.24
Specifications
Melting Point 132°C to 134°C
Density 1.31 g/cm3 at 20°C
Boiling Point 344°C/1023 hPa
MDL Number MFCD00004496
Quantity 1 kg
Synonym Benzoin, α-Hydroxy-α-phenylacetophenone, Benzoin