Learn More
Benzo[b]furan-2-carboxaldehyde, 99%
Product Code. 11317715
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
Product Code. Quantity unitSize
11317715 2 g 2g
11327715 10 g 10g
2 options
Product Code. 11317715
Brand: Thermo Scientific Alfa Aesar L02456.04
Request Bulk or Custom Format
CAS: 4265-16-1 | C9H6O2 | 146.145 g/mol
Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzo[b]furan-2-carboxaldehyde is used for the synthesis of isoxazolines.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4265-16-1
Molecular Formula C9H6O2
Molecular Weight (g/mol) 146.145
MDL Number MFCD00015463
InChI Key ADDZHRRCUWNSCS-UHFFFAOYSA-N
Synonym benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
PubChem CID 61341
IUPAC Name 1-benzofuran-2-carbaldehyde
SMILES C1=CC=C2C(=C1)C=C(O2)C=O
Specifications
Boiling Point 92°C to 97°C (1 mmHg)
Refractive Index 1.632
Quantity 2 g
Beilstein 2910
Sensitivity Air sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 146.15
Percent Purity 99%
Chemical Name or Material Benzo[b]furan-2-carboxaldehyde