Learn More
Benzenesulfonamide, Sigma-Aldrich™
Product Code. 15814651
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15814651 500 g 500g
15804651 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15814651 Supplier Sigma Aldrich Supplier No. 8219590500
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-10-2
Grade Research
Molecular Weight (g/mol) 157.19
Specifications
Melting Point 149°C to 152°C
MDL Number MFCD00007930
Quantity 500 g
Synonym Benzenesulfonamide, Benzenesulfonamide