Learn More
Bentazon, TRC
Product Code. 17911775
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911775 1 g 1g
17923655 5 g 5g
17930715 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17911775 Supplier Toronto Research Chemicals Supplier No. TRCB1205801G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25057-89-0
Molecular Formula C10 H12 N2 O3 S
Molecular Weight (g/mol) 240.28
Synonym 1H-2,1,3-Benzothiadiazin-4(3H)-one, 3-isopropyl-, 2,2-dioxide (8CI), 3,4-Dihydro-3-isopropyl-1H-2,1,3-benzothiadiazin-4-one 2,2-dioxide, 3,4-Dihydro-3-isopropyl-4-oxo-1H-2,1,3-benzothiadiazine 2,2-dioxide, 3-Isopropyl-1H-2,1,3-benzothiadiazin-4(3H)-one 2,2-dioxide, 3-Isopropyl-1H-2,1,3-benzothiadiazin-4(3H)-one S,S-dioxide, 3-Isopropyl-1H-2,1,3-benzothiadiazine-4(3H)-one 2,2-dioxide, 3-Isopropyl-2,1,3-benzothiadiazin-4-one 2,2-dioxide, 3-Isopropyl-2,1,3-benzothiadiazine-4-one 2,2-dioxide, 3-Isopropyl-2,1,3-benzothiadiazinon-(4)-2,2-dioxide, 3-Isopropyl-2,1,3-benzothiodiazin-4-one 2,2-dioxide, 3-Isopropyl-4-oxo-2,1,3-benzothiadiazine 2,2-dioxide, BAS 351-07H, BAS 3510, BAS 3510H, BAS 3512H, BAS 3517H, BAS 351H, Basagran, Basagran 480, Basagran 480SL, Basamais, Bazargan, Bendioxide, Bentazon, Bentazone, Corsar, Leader
IUPAC Name 2,2-dioxo-3-propan-2-yl-1H-2λ^{6},1,3-benzothiadiazin-4-one
SMILES CC(C)N1C(=O)c2ccccc2NS1(=O)=O
Specifications
Analyte or Component Names Bentazone
Shipping Condition Room Temperature
InChI Formula InChI=1S/C10H12N2O3S/c1-7(2)12-10(13)8-5-3-4-6-9(8)11-16(12,14)15/h3-7,11H,1-2H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 1 g
Formula Weight 240.0569
Chemical Name or Material Bentazone