Learn More
Corning™ Bent Discharge Tubes
Product Code. 16882120 Shop All Corning Products
Click to view available options
Volume (Metric):
2.5 to 10 mL
25 to 100 mL
Unit Size:
Each
Product Code. 16882120
Brand: Corning™ 6879
Corning™ Bent Discharge Tubes for quick and precise dispensing of acids, bases and solvents
  • Bent
  • Includes protective sleeve
TRUSTED_SUSTAINABILITY
Specifications
Angle Bent
Product Type Discharge Tube
Material FEP, Polypropylene
Volume (Metric) 2.5 to 10 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) 2.5 to 10 mL Bottle Top Dispenser
Includes Protective Sleeve
Length (Metric) 80 mm