Learn More
Lenz Laborglasinstrumente™ Bent Adapter
Product Code. 11896154
Change view
Click to view available options
Standard Taper Joint:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 29/32 Upper Socket, 29/32 NS Lower Socket
Type:
Bent Cone Adapter
Bent Socket Adapter
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Type Unit Size
11896154 NS 24/29 Bent Cone Adapter Each
11816084 NS 14/23 Bent Cone Adapter Each
11856174 NS 29/32 Bent Socket Adapter Each
11876144 NS 14/23 Bent Socket Adapter Each
11856194 NS 19/26 Bent Cone Adapter Each
10740232 NS 29/32 Upper Socket, 29/32 NS Lower Socket Bent Socket Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11896154 Supplier Lenz Laborglasinstrumente™ Supplier No. 05095003
  • ONLINE EXCLUSIVE PRICE
    194.00 NOK
    162.15 NOK / Pack of 1
    Save 31.85 NOK
    16% Off
Estimated Shipment: 11-12-2026
Allows for tubing connections. Lenz Laborglasinstrumente™ Bent Adapters feature a bent glass olive at 90°.
  • Constructed from DURAN™ tubing
  • With NS socket
  • Bore: 2.5 mm
Specifications
Type Bent Cone Adapter
Material Duran Glass
Standard Taper Joint NS 24/29