missing translation for 'onlineSavingsMsg'
Learn More

Benserazide Hydrochloride, TRC

Product Code. 17944945
Change view
Click to view available options
Quantity:
10 mg
100 mg
5 g
Unit Size:
100mg
10mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944945 10 mg 10mg
30135540 100 mg 100mg
30135898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944945 Supplier Toronto Research Chemicals Supplier No. TRCB12050010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14919-77-8
Formula Weight 293.0778
InChI Formula InChI=1S/C10H15N3O5.ClH/c11-6(4-14)10(18)13-12-3-5-1-2-7(15)9(17)8(5)16;/h1-2,6,12,14-17H,3-4,11H2,(H,13,18);1H
IUPAC Name 2-amino-3-hydroxy-N'-[(2,3,4-trihydroxyphenyl)methyl]propanehydrazide;hydrochloride
Molecular Formula C10 H15 N3 O5 . Cl H
Molecular Weight (g/mol) 293.7
SMILES Cl.NC(CO)C(=O)NNCc1ccc(O)c(O)c1O
Synonym Benserazide hydrochloride, (2RS)-2-Amino-3-hydroxy-2'-(2,3,4-trihydroxybenzyl)propanohydrazide hydrochloride
Chemical Name or Material Benserazide Hydrochloride

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.