Learn More
Chem-Lab Benedict's reagent (173g 2Na3C
Brand:  Chem-Lab CL02.0214.1000
Benedict's reagent (173g 2Na3C
Product Code. 13288293
  • 1420.00 NOK / 1 L
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium carbonate