Learn More
Thermo Scientific™ Benchtop Dewar Flasks
Beskrivning
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.
- High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
- Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
- Use from -196°C to +100°C
- Pour spout and bottom grip area for ease of removal of used liquids
- Vented, insulated cover allows for vapor escape but maintains sample temperature
Specifikationer
Specifikationer
| Capacity (English) | 338.14 fl oz. |
| No. per Case | 1 |
| No. per Pack | 1 |
| Capacity (Metric) | 10 L |
| Depth (Metric) Interior | 396 mm |
| Depth (English) Interior | 15.6 in. |
| Diameter (Metric) Inner Mouth | 198mm |
| Diameter (English) Inner Mouth | 7.8 in. |
| Height (Metric) | 470mm |
| Height (English) | 18.5 in. |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?