Learn More
Koettermann™ Bench with Wheels
Product Code. 15508585
Change view
Click to view available options
:
Height: 420mm
Product Code.
15508585 Height: 420mm
1 options
Product Code. 15508585
Brand: Koettermann™ 94500003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Koettermann™ Bench with Wheels is mobile and suitable for laboratory conditions.
  • Mobile bench
  • Wheels
Specifications
Product Type Mobile Bench
Color Gray
Height (Metric) 420 mm