Learn More
Kern Sohn Bench Scale FCE
Product Code. 17381633
Encompass_Preferred
Change view
Click to view available options
Capacity:
3 kg
6 kg
15 kg
30 kg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
17381633 6 kg Each
17341633 15 kg Each
17351633 30 kg Each
17361633 3 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17381633 Supplier Kern Sohn Supplier No. FCE 6K2N
Estimated Shipment: 13-11-2026
Entry-level bench scale that is mobile, practical, and lightweight.
  • Simple and convenient 2-key operation
  • Very fast display: steady weight values within 3 s
  • Ideal as a compact letter and parcel scale, especially where space is limited
  • Ideal as a grading, commissioning or simple check-weighing scale in production or dispatch
  • Protective working cover included with delivery
Specifications
Capacity 6 kg
Dimensions (LxW) Platform 228 x 252 mm
Repeatability 2 g