Learn More
Kern Sohn Bench Scale FCE
Product Code. 17341633
Encompass_Preferred
Click to view available options
Capacity:
3 kg
6 kg
15 kg
30 kg
Unit Size:
Each
Product Code. 17341633
Brand: Kern Sohn FCE 15K5N
Entry-level bench scale that is mobile, practical, and lightweight.
  • Simple and convenient 2-key operation
  • Very fast display: steady weight values within 3 s
  • Ideal as a compact letter and parcel scale, especially where space is limited
  • Ideal as a grading, commissioning or simple check-weighing scale in production or dispatch
  • Protective working cover included with delivery
Specifications
Capacity 15 kg
Dimensions (LxW) Platform 228 x 252 mm
Repeatability 10 g
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.