Learn More
Bempedoic acid, MedChemExpress
Product Code. 16485118
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16485118
Brand: Medchem Express HY12357
MedChemExpress Bempedoic acid (ETC-1002) is an ATP-citrate lyase (ACL) inhibitor. Bempedoic acid (ETC-1002) activates AMPK.
Bempedoic acid (ETC-1002) is an ATP-citrate lyase (ACL) inhibitor. Bempedoic acid (ETC-1002) activates AMPK.
Specifications
Chemical Name or Material Bempedoic acid
Physical Form Solid
CAS 738606-46-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H36O5
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym ETC-1002 ESP-55016
Solubility Information DMSO : 100 mg/mL (290.28 mM; Need ultrasonic)
SMILES O=C(O)C(C)(C)CCCCCC(O)CCCCCC(C)(C)C(O)=O
Molecular Weight (g/mol) 344.49
Formula Weight 344.49
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.